Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ABS 037-35-02 detects aprotinin in bovine samples.
ABS 037-35-02 has successfully been used in ELISA procedures. PBS buffers are unsuitable for this assay and 0.2M glycine sodium buffer, pH 8.8, is recommended for washing and dilution buffers.
Aprotinin, also called bovine pancreatic trypsin inhibitor, is a single chain globular Kunitz-type serine proteinase inhibitor found in bovine pancreas, parotid gland and lung. It forms stable, enzymatically inactive complexes with trypsin, chymotrypsin, plasmin and kallikrein. Aprotinin has been used therapeutically as an antifibrinolytic agent under the name Trasylol (Bayer) to reduce blood loss during cardiopulmonary bypass surgery but this use has been associated with an increased frequency of acute renal injury.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aprotinin; basic protease inhibitor; trypsin ihibitor
Gene Aliases: BPI; BPTI
Entrez Gene ID: (Bovine) 404172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support